1-[2-(Allyloxy)benzyl]piperazine

Supplier: Matrix Scientific

029382-500MG
101828-036EA 290.77 USD
101828-036
1-[2-(Allyloxy)benzyl]piperazine
1-[2-(Allyloxy)benzyl]piperazine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MDL Number: MFCD09434010
CAS Number: 925650-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR