N,N-Dimethyl-N-[4-(piperazin-1-ylmethyl)phenyl]amine

Supplier: Matrix Scientific

029381-500MG
101828-034EA 217.35 USD
101828-034
N,N-Dimethyl-N-[4-(piperazin-1-ylmethyl)phenyl]amine
N,N-Dimethyl-N-[4-(piperazin-1-ylmethyl)phenyl]amine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD03069505
CAS Number: 89292-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR