1-[(5-Ethylthien-2-yl)methyl]piperazine

Supplier: Matrix Scientific

029380-500MG
101828-032EA 290.77 USD
101828-032
1-[(5-Ethylthien-2-yl)methyl]piperazine
1-[(5-Ethylthien-2-yl)methyl]piperazine

Hazard: Irritant

Formula: C₁₁H₁₈N₂S
MDL Number: MFCD05189251
CAS Number: 523981-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR