1-[(5-Chlorothien-2-yl)methyl]piperazine

Supplier: Matrix Scientific

029378-500MG
101828-028EA 540 USD
101828-028
1-[(5-Chlorothien-2-yl)methyl]piperazine
1-[(5-Chlorothien-2-yl)methyl]piperazine

Hazard: Irritant

Formula: C₉H₁₃ClN₂S
MDL Number: MFCD06370793
CAS Number: 55513-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR