1-(2-Ethylbutyl)piperazine

Supplier: Matrix Scientific

029376-500MG
101828-024EA 217.35 USD
101828-024
1-(2-Ethylbutyl)piperazine
1-(2-Ethylbutyl)piperazine

Hazard: Irritant

Formula: C₁₀H₂₂N₂
MDL Number: MFCD09434008
CAS Number: 57184-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR