1-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029374-500MG
101828-020EA 290.77 USD
101828-020
1-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₃ClFN₃
MDL Number: MFCD02090912
CAS Number: 512809-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR