1-(2-Chloro-4-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029373-500MG
101828-018EA 290.77 USD
101828-018
1-(2-Chloro-4-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(2-Chloro-4-fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₃ClFN₃
MDL Number: MFCD04967280
CAS Number: 925663-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR