1-(2,6-Dichlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029370-500MG
101828-012EA 290.77 USD
101828-012
1-(2,6-Dichlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(2,6-Dichlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₃Cl₂N₃
MDL Number: MFCD02055613
CAS Number: 898055-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR