1-(3-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029368-500MG
101828-008EA 290.77 USD
101828-008
1-(3-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(3-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₄FN₃
MDL Number: MFCD03422518
CAS Number: 925650-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR