1-(2-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029367-500MG
101828-006EA 290.77 USD
101828-006
1-(2-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(2-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₃
MDL Number: MFCD02055747
CAS Number: 514801-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR