1-(3-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029366-500MG
101828-004EA 290.77 USD
101828-004
1-(3-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(3-Chlorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₃
MDL Number: MFCD04969191
CAS Number: 925663-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR