3,5-Dimethyl-1-(3-methylbenzyl)-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029362-500MG
101827-996EA 290.77 USD
101827-996
3,5-Dimethyl-1-(3-methylbenzyl)-1H-pyrazol-4-amine
3,5-Dimethyl-1-(3-methylbenzyl)-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₃H₁₇N₃
MDL Number: MFCD04967275
CAS Number: 955964-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR