1-Benzyl-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

029360-500MG
101827-992EA 165.6 USD
101827-992
1-Benzyl-3,5-dimethyl-1H-pyrazol-4-amine
1-Benzyl-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₅N₃
MDL Number: MFCD01693691
CAS Number: 28466-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR