1-(3-Isopropoxyphenyl)ethanamine

Supplier: Matrix Scientific

029359-500MG
101827-990EA 129.95 USD
101827-990
1-(3-Isopropoxyphenyl)ethanamine
1-(3-Isopropoxyphenyl)ethanamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD09434007
CAS Number: 854304-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR