1-(3-Propoxyphenyl)ethanamine

Supplier: Matrix Scientific

029358-500MG
101827-988EA 338.46 USD
101827-988
1-(3-Propoxyphenyl)ethanamine
1-(3-Propoxyphenyl)ethanamine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD09434006
CAS Number: 925650-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR