1-(3-Butoxyphenyl)ethanamine

Supplier: Matrix Scientific

029357-500MG
101827-986EA 144.9 USD
101827-986
1-(3-Butoxyphenyl)ethanamine
1-(3-Butoxyphenyl)ethanamine

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD09431259
CAS Number: 925180-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR