1-(3-Ethoxyphenyl)ethanamine

Supplier: Matrix Scientific

029353-500MG
101827-978EA 129.23 USD
101827-978
1-(3-Ethoxyphenyl)ethanamine
1-(3-Ethoxyphenyl)ethanamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD05215235
CAS Number: 603945-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR