1-(4-Ethoxyphenyl)propan-1-amine

Supplier: Matrix Scientific

029351-500MG
101827-976EA 193.85 USD
101827-976
1-(4-Ethoxyphenyl)propan-1-amine
1-(4-Ethoxyphenyl)propan-1-amine

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD09431258
CAS Number: 925608-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR