1-(4-Chlorophenyl)propan-1-amine

Supplier: Matrix Scientific

029350-500MG
101827-974EA 307.69 USD
101827-974
1-(4-Chlorophenyl)propan-1-amine
1-(4-Chlorophenyl)propan-1-amine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD08691240
CAS Number: 74788-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR