1-(3-Chlorophenyl)propan-1-amine

Supplier: Matrix Scientific

029349-500MG
101827-972EA 407.69 USD
101827-972
1-(3-Chlorophenyl)propan-1-amine
1-(3-Chlorophenyl)propan-1-amine

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD09431257
CAS Number: 40023-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR