1-(2,4-Dimethylphenyl)propan-1-amine

Supplier: Matrix Scientific

029347-500MG
101827-968EA 323.08 USD
101827-968
1-(2,4-Dimethylphenyl)propan-1-amine
1-(2,4-Dimethylphenyl)propan-1-amine

Hazard: Irritant

Formula: C₁₁H₁₇N
MDL Number: MFCD06739390
CAS Number: 886496-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR