1-(2,4-Dichlorophenyl)ethylamine

Supplier: Matrix Scientific

Synonyms: 1-Amino-1-(2,4-dichlorophenyl)ethane2,4-Dichloro-alpha-methylbenzylamine1-(2,4-Dichlorophenyl)ethanamine

029346-500MG
101827-966EA 15.38 USD
101827-966
1-(2,4-Dichlorophenyl)ethylamine
1-(2,4-Dichlorophenyl)ethylamine

Hazard: Irritant

Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
MDL Number: MFCD05215236
CAS Number: 89981-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR