2-tert-Butylcyclohexanamine

Supplier: Matrix Scientific

029345-500MG
101827-964EA 252.31 USD
101827-964
2-tert-Butylcyclohexanamine
2-tert-Butylcyclohexanamine

Hazard: Irritant

Formula: C₁₀H₂₁N
MDL Number: MFCD08558493
CAS Number: 35735-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR