4-[(2-Ethylpiperidin-1-yl)sulfonyl]aniline

Supplier: Matrix Scientific

029344-500MG
101827-962EA 193.85 USD
101827-962
4-[(2-Ethylpiperidin-1-yl)sulfonyl]aniline
4-[(2-Ethylpiperidin-1-yl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD03050282
CAS Number: 436091-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR