4-Amino-N-isobutylbenzenesulfonamide

Supplier: Matrix Scientific

029343-500MG
101827-960EA 252.31 USD
101827-960
4-Amino-N-isobutylbenzenesulfonamide
4-Amino-N-isobutylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MDL Number: MFCD03753719
CAS Number: 53668-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR