4-Amino-N-(1-phenylethyl)benzenesulfonamide

Supplier: Matrix Scientific

029340-500MG
101827-956EA 193.85 USD
101827-956
4-Amino-N-(1-phenylethyl)benzenesulfonamide
4-Amino-N-(1-phenylethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD00544446
CAS Number: 79867-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR