Ethyl-5-amino-3-methylisothiazole-4-carboxylate

Supplier: Matrix Scientific

029339-500MG
101827-954EA 134.55 USD
101827-954
Ethyl-5-amino-3-methylisothiazole-4-carboxylate
Ethyl-5-amino-3-methylisothiazole-4-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD00097916
CAS Number: 34859-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR