4-(3,4-Dihydroisoquinolin-2(1H)-ylsulfonyl)aniline

Supplier: Matrix Scientific

029338-500MG
101827-952EA 193.85 USD
101827-952
4-(3,4-Dihydroisoquinolin-2(1H)-ylsulfonyl)aniline
4-(3,4-Dihydroisoquinolin-2(1H)-ylsulfonyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂S
MDL Number: MFCD06761407
CAS Number: 7252-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR