4-[(6-Fluoro-2-methyl-3,4-dihydroquinolin-1(2H)-yl)sulfonyl]aniline

Supplier: Matrix Scientific

029337-500MG
101827-950EA 144.9 USD
101827-950
4-[(6-Fluoro-2-methyl-3,4-dihydroquinolin-1(2H)-yl)sulfonyl]aniline
4-[(6-Fluoro-2-methyl-3,4-dihydroquinolin-1(2H)-yl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₇FN₂O₂S
MDL Number: MFCD06757626
CAS Number: 774586-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR