1-[(5-Bromothien-2-yl)sulfonyl]piperazine

Supplier: Matrix Scientific

029335-500MG
101827-946EA 144.9 USD
101827-946
1-[(5-Bromothien-2-yl)sulfonyl]piperazine
1-[(5-Bromothien-2-yl)sulfonyl]piperazine

Hazard: Irritant

Formula: C₈H₁₁BrN₂O₂S₂
MDL Number: MFCD05664870
CAS Number: 725688-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR