1-[(4-tert-Butylphenyl)sulfonyl]piperazine

Supplier: Matrix Scientific

029324-500MG
101827-924EA 193.85 USD
101827-924
1-[(4-tert-Butylphenyl)sulfonyl]piperazine
1-[(4-tert-Butylphenyl)sulfonyl]piperazine

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂S
MDL Number: MFCD02655039
CAS Number: 379244-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR