1-[(4-Isopropylphenyl)sulfonyl]piperazine

Supplier: Matrix Scientific

029323-500MG
101827-922EA 193.85 USD
101827-922
1-[(4-Isopropylphenyl)sulfonyl]piperazine
1-[(4-Isopropylphenyl)sulfonyl]piperazine

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD03444528
CAS Number: 730976-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR