4-[(2,6-Dimethylpiperidin-1-yl)sulfonyl]aniline

Supplier: Matrix Scientific

029320-500MG
101827-916EA 193.85 USD
101827-916
4-[(2,6-Dimethylpiperidin-1-yl)sulfonyl]aniline
4-[(2,6-Dimethylpiperidin-1-yl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD02018979
CAS Number: 345991-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR