2-Amino-4,5-dihydronaphtho[1,2-b]thiophene-3-carboxamide

Supplier: Matrix Scientific

029317-500MG
101827-910EA 233.85 USD
101827-910
2-Amino-4,5-dihydronaphtho[1,2-b]thiophene-3-carboxamide
2-Amino-4,5-dihydronaphtho[1,2-b]thiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₂N₂OS
MDL Number: MFCD04055510
CAS Number: 617688-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR