Diethyl-5-amino-3-propylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

029314-500MG
101827-906EA 290.77 USD
101827-906
Diethyl-5-amino-3-propylthiophene-2,4-dicarboxylate
Diethyl-5-amino-3-propylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₉NO₄S
MDL Number: MFCD03900731
CAS Number: 667437-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR