2-Amino-N-cyclopropylthiophene-3-carboxamide

Supplier: Matrix Scientific

029309-500MG
101827-896EA 259.9 USD
101827-896
2-Amino-N-cyclopropylthiophene-3-carboxamide
2-Amino-N-cyclopropylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₈H₁₀N₂OS
MDL Number: MFCD03945607
CAS Number: 590352-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR