2-Amino-N-(2-furylmethyl)thiophene-3-carboxamide

Supplier: Matrix Scientific

029298-500MG
101827-874EA 217.35 USD
101827-874
2-Amino-N-(2-furylmethyl)thiophene-3-carboxamide
2-Amino-N-(2-furylmethyl)thiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MDL Number: MFCD03944472
CAS Number: 588693-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR