Methyl-2-{[(2-aminothien-3-yl)carbonyl]amino}thiophene-3-carboxylate

Supplier: Matrix Scientific

029296-500MG
101827-870EA 217.35 USD
101827-870
Methyl-2-{[(2-aminothien-3-yl)carbonyl]amino}thiophene-3-carboxylate
Methyl-2-{[(2-aminothien-3-yl)carbonyl]amino}thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃S₂
MDL Number: MFCD03946008
CAS Number: 590356-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR