2-Amino-4-ethyl-5-methyl-N-phenylthiophene-3-carboxamide

Supplier: Matrix Scientific

029295-500MG
101827-868EA 193.85 USD
101827-868
2-Amino-4-ethyl-5-methyl-N-phenylthiophene-3-carboxamide
2-Amino-4-ethyl-5-methyl-N-phenylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂OS
MDL Number: MFCD03945730
CAS Number: 590353-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR