2-Amino-N-(2,4-dichlorophenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Supplier: Matrix Scientific

029288-500MG
101827-854EA 144.9 USD
101827-854
2-Amino-N-(2,4-dichlorophenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
2-Amino-N-(2,4-dichlorophenyl)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₄Cl₂N₂OS
MDL Number: MFCD03945430
CAS Number: 590350-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR