6-tert-Butyl-3-(morpholin-4-ylcarbonyl)-4,5,6,7-tetrahydro-1-benzothien-2-ylamine

Supplier: Matrix Scientific

029286-500MG
101827-850EA 144.9 USD
101827-850
6-tert-Butyl-3-(morpholin-4-ylcarbonyl)-4,5,6,7-tetrahydro-1-benzothien-2-ylamine
6-tert-Butyl-3-(morpholin-4-ylcarbonyl)-4,5,6,7-tetrahydro-1-benzothien-2-ylamine

Hazard: Irritant

Formula: C₁₇H₂₆N₂O₂S
MDL Number: MFCD03945474
CAS Number: 590351-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR