2-Amino-N-cyclopropyl-4-(4-methylphenyl)thiophene-3-carboxamide

Supplier: Matrix Scientific

029285-500MG
101827-848EA 193.85 USD
101827-848
2-Amino-N-cyclopropyl-4-(4-methylphenyl)thiophene-3-carboxamide
2-Amino-N-cyclopropyl-4-(4-methylphenyl)thiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂OS
MDL Number: MFCD03946005
CAS Number: 590356-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR