2-Amino-N-(2,5-dichlorophenyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Supplier: Matrix Scientific

029284-500MG
101827-846EA 144.9 USD
101827-846
2-Amino-N-(2,5-dichlorophenyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
2-Amino-N-(2,5-dichlorophenyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₆H₁₆Cl₂N₂OS
MDL Number: MFCD03944687
CAS Number: 588696-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR