5-Oxo-5-[3-oxo-2-(2-oxo-2-propoxyethyl)-1-piperazinyl]pentanoic acid

Supplier: Matrix Scientific

024018-500MG
101817-238EA 228.85 USD
101817-238
5-Oxo-5-[3-oxo-2-(2-oxo-2-propoxyethyl)-1-piperazinyl]pentanoic acid
5-Oxo-5-[3-oxo-2-(2-oxo-2-propoxyethyl)-1-piperazinyl]pentanoic acid

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₆
MDL Number: MFCD05881342
CAS Number: 1024712-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR