Propyl-4-(2-hydrazino-2-oxoethoxy)benzoate

Supplier: Matrix Scientific

024014-500MG
101817-230EA 96.6 USD
101817-230
Propyl-4-(2-hydrazino-2-oxoethoxy)benzoate
Propyl-4-(2-hydrazino-2-oxoethoxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD06447102
CAS Number: 847468-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR