2-(2-Bromo-4-ethylphenoxy)acetohydrazide

Supplier: Matrix Scientific

024001-500MG
101817-204EA 228.85 USD
101817-204
2-(2-Bromo-4-ethylphenoxy)acetohydrazide
2-(2-Bromo-4-ethylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O₂
MDL Number: MFCD03488728
CAS Number: 496029-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR