2-(4-Bromo-2-methylphenoxy)acetohydrazide

Supplier: Matrix Scientific

023999-500MG
101817-200EA 228.85 USD
101817-200
2-(4-Bromo-2-methylphenoxy)acetohydrazide
2-(4-Bromo-2-methylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁BrN₂O₂
MDL Number: MFCD01608855
CAS Number: 401611-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR