N-[4-(Hydrazinocarbonyl)phenyl]cyclopropanecarboxamide

Supplier: Matrix Scientific

023989-500MG
101817-180EA 134.55 USD
101817-180
N-[4-(Hydrazinocarbonyl)phenyl]cyclopropanecarboxamide
N-[4-(Hydrazinocarbonyl)phenyl]cyclopropanecarboxamide

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MDL Number: MFCD03109882
CAS Number: 446278-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR