2-(2,4-Dibromo-6-methylphenoxy)acetohydrazide

Supplier: Matrix Scientific

023986-500MG
101817-174EA 228.85 USD
101817-174
2-(2,4-Dibromo-6-methylphenoxy)acetohydrazide
2-(2,4-Dibromo-6-methylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₀Br₂N₂O₂
MDL Number: MFCD02604498
CAS Number: 356100-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR