2-(2,4-Dibromophenoxy)acetohydrazide

Supplier: Matrix Scientific

023984-500MG
101817-170EA 113.85 USD
101817-170
2-(2,4-Dibromophenoxy)acetohydrazide
2-(2,4-Dibromophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₈Br₂N₂O₂
MDL Number: MFCD00098698
CAS Number: 141244-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR